Learn More
Thermo Scientific™ Cover for Precision™ Water Baths
Shop All Thermo Scientific Products
Click to view available options
Type:
Gable Cover
Large Gassing Hood
Small Gassing Hood
Material:
Polycarbonate
Stainless Steel
For Use With (Equipment):
Precision 10L Water Baths
Precision 15L Shaking Water Baths
Precision 20L and 28L Water Baths
Precision 2L Water Bath
Precision 2L and 5L Water Baths
Unit Size:
Each
Description
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods
- Permit control over the atmosphere surrounding the sample
- Improve temperature uniformity and reduce energy consumption
- Polished stainless steel with plastic gas fitting
- Each bath will accommodate 1 large hood or 2 small hoods
Warranty
1 year
Specifications
Specifications
| For Use With (Equipment) | Precision 15L Shaking Water Baths |
| Type | Large Gassing Hood |
| Material | Stainless Steel |