missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gasoline Range Organics (EPA), SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep GRO-EPA
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,2,4-Trimethylbenzene | 95-63-6 | 1500μg/mL |
2,2,4-Trimethylpentane | 540-84-1 | 1500μg/mL |
2-Methylpentane | 107-83-5 | 1500μg/mL |
m-Xylene | 108-38-3 | 1500μg/mL |
o-Xylene | 95-47-6 | 1500μg/mL |
Toluene | 108-88-3 | 1500μg/mL |
Benzene | 71-43-2 | 500μg/mL |
Ethylbenzene | 100-41-4 | 500μg/mL |
n-Heptane | 142-82-5 | 500μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Gasoline Range Organics (EPA) | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
Methanol-P&T |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Gasoline Range Organics (EPA), Alternate Form
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol, toluene
Product intended for laboratory use only