Learn More
Usbeck Laborgeraete™ Gas Lighters
Brand:  Usbeck Laborgeraete™ 1650
 View more versions of this product
Product Code. 10779542
  • € 15.75 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Specifications
220 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Usbeck Laborgeraete™ Gas Lighters > Length: 220mm