Learn More
Ansell™ Gammex™ PF Powder-Free Latex Surgical Gloves
Product Code. 13246444
Click to view available options
Size:
7.5
9
Quantity:
40 pairs per box; 6 boxes per case
50 Pairs/Box
Unit Size:
40 pairs
50 pairs
Pack of 40
Product Code. 13246444
Brand: Ansell™ 351148
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides an optimal combination of safety, based on Ansell's powder-free technology, with paralleled levels of comfort and ease-of-use. Ansell Edmont™ Gammex™ PF Powder-Free Latex Surgical Gloves have a very high tear resistance, with a favorable strength-to-thickness ratio for optimal barrier protection.
  • Micro-rough outer surface provides a secure grip in both wet and dry conditions
  • Synthetic inner coating reduces possible contact with latex allergens
  • Resistant to alcohol-based hand disinfectants and viral penetration
  • Delivers elasticity, sensitivity and comfort plus a natural fit that helps prevent hand fatigue
Specifications
Disposable Single-use
Hand Ambidextrous
Latex Free No
Material Latex
Powdered Powder Free
Size 9
Glove Cuff Style Straight
Quantity 40 pairs per box; 6 boxes per case
Resistant To Tears, alcohol-based hand disinfectants, viral penetration