Learn More
Ansell™ Gammex™ PF Powder-Free Latex Surgical Gloves
Provides an optimal combination of safety, based on Ansell's powder-free technology, with paralleled levels of comfort and ease-of-use. Ansell Edmont™ Gammex™ PF Powder-Free Latex Surgical Gloves have a very high tear resistance, with a favorable strength-to-thickness ratio for optimal barrier protection.
Brand:  Ansell™ 351145
 View more versions of this product
Product Code. 13236444
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Micro-rough outer surface provides a secure grip in both wet and dry conditions
  • Synthetic inner coating reduces possible contact with latex allergens
  • Resistant to alcohol-based hand disinfectants and viral penetration
  • Delivers elasticity, sensitivity and comfort plus a natural fit that helps prevent hand fatigue
Specifications
Specifications
Single-use
No
Powder Free
Straight
Tears, alcohol-based hand disinfectants, viral penetration
Ambidextrous
Latex
7.5
40 pairs per box; 6 boxes per case