missing translation for 'onlineSavingsMsg'
Learn More

gamma-Valerolactone, 98%, Thermo Scientific Chemicals

€ 27.14 - € 240.20

Chemical Identifiers

CAS 108-29-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00005400
InChI Key GAEKPEKOJKCEMS-UHFFFAOYSA-N
Synonym gamma-valerolactone, 4-valerolactone, 4-pentanolide, 5-methyldihydrofuran-2 3h-one, 4-hydroxypentanoic acid lactone, gamma-pentalactone, 4-hydroxyvaleric acid lactone, 2 3h-furanone, dihydro-5-methyl, 4-methyl-gamma-butyrolactone, pentanolide-1,4
PubChem CID 7921
ChEBI CHEBI:48569
IUPAC Name 5-methyloxolan-2-one
SMILES CC1CCC(=O)O1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11458900
View Documents
Thermo Scientific Alfa Aesar
A18872.14
25 g
€ 27.14
25g
Please to purchase this item. Need a web account? Register with us today!
11413128
View Documents
Thermo Scientific Alfa Aesar
A18872.22
100 g
€ 58.99
100g
Please to purchase this item. Need a web account? Register with us today!
11423128
View Documents
Thermo Scientific Alfa Aesar
A18872.36
500 g
€ 240.20
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It finds it application as a food additive that is used to improve the taste or odor of a food. It is also used as toiletry fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a food additive that is used to improve the taste or odor of a food. It is also used as toiletry fragrances.

Solubility
Fully miscible in water (9.381e+004 mg/L @ 25°C (est.)). Soluble in alcohol, fixed oils.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

108-29-2
100.117
GAEKPEKOJKCEMS-UHFFFAOYSA-N
7921
5-methyloxolan-2-one
C5H8O2
MFCD00005400
gamma-valerolactone, 4-valerolactone, 4-pentanolide, 5-methyldihydrofuran-2 3h-one, 4-hydroxypentanoic acid lactone, gamma-pentalactone, 4-hydroxyvaleric acid lactone, 2 3h-furanone, dihydro-5-methyl, 4-methyl-gamma-butyrolactone, pentanolide-1,4
CHEBI:48569
CC1CCC(=O)O1

Specifications

108-29-2
1.052
81°C (178°F)
C5H8O2
MFCD00005400
gamma-valerolactone, 4-valerolactone, 4-pentanolide, 5-methyldihydrofuran-2 3h-one, 4-hydroxypentanoic acid lactone, gamma-pentalactone, 4-hydroxyvaleric acid lactone, 2 3h-furanone, dihydro-5-methyl, 4-methyl-gamma-butyrolactone, pentanolide-1,4
GAEKPEKOJKCEMS-UHFFFAOYSA-N
5-methyloxolan-2-one
7921
100.12
gamma-Valerolactone
-31°C
207°C to 208°C
Pleasant
1.433
80420
Fully miscible in water (9.381e+004mg/L @ 25°C (est.)). Soluble in alcohol,fixed oils.
CC1CCC(=O)O1
100.117
CHEBI:48569
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 203-569-5

missing translation for 'rtecsNumber' : LU3580000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only