Learn More
Fisherbrand™ Gamma Radiation Safe Transport Box
Provides radiation protection shielding for use in laboratories.
Brand: Fisherbrand™ CSR-B8G
Description
- Manufactured with hinged lids
- Transparent shielding
- Serves as solution for short-term storage of radioactive waste or radioisotopes
- Constructed of acrylic provides easy fabrication and manufacturability
Specifications
29.5 x 38 x 7.5cm (11.61 x 14.96 x 2.95 in.) | |
Radioisotopes or radioactive waste | |
Transport Box |
Gamma radiation shielded | |
Hinged lid | |
Acrylic |
Product Suggestions
Customers who viewed this item also viewed.