Learn More
SCAT™ Safety Funnel 'ARNOLD' with Ball and Lid, V2.0, HDPE-EC
Product Code. 17112235 Shop All SCAT Europe Products
Click to view available options
Thread Size (Metric):
B83
GL45
R2
S50
S51
S55
S60/61
S65
S70/71
S90
S95
Includes:
Splash Guard, 2 x Removable Sieves, Grounding Cable
Splash Guard, Sieve, Grounding Cable
Splash Guard, Sieve, Grounding Cable, Ball Valve
Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance
For Use With (Equipment):
Disposal Containers
High Flow Solution System
Unit Size:
Each
Product Code. 17112235
Brand: SCAT™ 317624
Only null left
Add to basket
Add to basket
The strong and electrically conductive safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.
The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.
  • Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
  • Electrically conductive HDPE material with grounding connection to prevent flying sparks
  • Hinged lid with sealing ring for closure when the funnel is not in use
  • Optimized sieve for collecting larger particles and for depositing laboratory bottles
  • Improved splash guard with integrated air channels to prevent bubble formation
  • Ball valve with automatic opening mechanism during filling
  • Individually shortenable safety lance for clean drainage of liquid waste
  • All major components are removable for easy cleaning
Specifications
Diameter (Metric)Top 200 mm (Funnel)
Capacity (Metric) 1500 mL
Material HDPE-EC
Height (Metric) 210 cm
Autoclavable Not Autoclavable
Color Black
For Use With (Application) Safe Disposal of Liquid and Flammable Wastes
For Use With (Equipment) Disposal Containers
Includes Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance
Diameter (English)Top 7.9 in. (Funnel)
Thread Size (Metric) S51
Product Type Safety Funnel with Hinged Lid