Learn More
Thermo Scientific™ Freezer and Refrigerator Shelves
Product Code. 11747754 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
23.3, 45.8, 51.1, 78.8 cu. ft. Units
29.2 cu. ft. Units
4.7 cu. ft., 11.5 cu. ft. Units
51.1 cu. ft. Units
Type:
Open Wire Shelves
Unit Size:
Each
Product Code. 11747754
Brand: Thermo Scientific™ 6697
Only null left
Add to basket
Add to basket
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
TRUSTED_SUSTAINABILITY
Specifications
Type Open Wire Shelves
Exterior Finish Epoxy-coated Steel
For Use With (Equipment) 29.2 cu. ft. Units
Warranty Warranty varies by country, please contact us for details
Holds General Freezer Items