Learn More
BUCHI Freeze Drying Beaker Flasks and Accessories
Freeze drying beaker flasks and accessories for use with a lyophilizer.
Brand: BUCHI 11065727
This item is not returnable.
View return policy
Description
- For use with a lyophilizer
Specifications
100 to 200 mL | |
L-200 | |
-60°C to +30°C | |
72 mm |
Freeze Drying | |
Beaker Flask Cap | |
0.6 lb. | |
0.3 kg |