missing translation for 'onlineSavingsMsg'
Learn More

FOXP4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 29311-1-AP-150UL

 View more versions of this product

Product Code. 17639656

  • € 410.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FOXP4 Polyclonal antibody apecifically detetects FOXP4 antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

FOXP4
Polyclonal
Unconjugated
BC052803
Rabbit
Antigen affinity purification
Primary
Human, Mouse
Liquid
Western Blot, Immunohistochemistry, ELISA
800 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
FOXP4
FOXP4 fusion protein Ag29953
150 μL
116113
Store at -20°C. Stable for one year after shipment.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.