Learn More
Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
10 to 50 mL
2 to 10 mL
missing translation for 'unitSize'
Each
Description
For dispensing of all chemicals - including solvents - that do not attack polypropylene and don't crystallize. Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers come with a glass piston.
- Visible dispensing procedure
- Easy to use volume setting system
- Valve block made of polypropylene with FIX adapter
Specifications
Specifications
| Accuracy | 0.60% |
| Product Type | Analog Bottletop Dispenser |
| Color | Natural |
| Channels | 1 |
| Material | PTFE, Polypropylene |
| Volume (Metric) | 10 to 50 mL |
| Increments | 1 mL |