Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringe
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
30 mL
50 mL
Unit Size:
Each
Description
- Complies with CE EN 46001 standards
- Constructed from Durobax™ glass
- With permanent amber graduation
- Acid and alkali resistant
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | CE EN 46001 |
| Material | Borosilicate Glass |
| Description | Amber graduation |
| Product Type | All Glass Luer-lock Syringe |
| Graduations | 0.2 mL |
| Quantity | 1/Ea. |
| Volume (Metric) | 5 mL |
| Connection Type | Luer Lock |
| Graduated | Yes |