Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringe
Product Code. 10252111
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
30 mL
50 mL
Unit Size:
Each
Product Code. 10252111
Brand: Poulten Graf™ 7 14037
Edge
Able to be autoclaved at 134°C. Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringes come with an interchangeable piston.
  • Complies with CE EN 46001 standards
  • Constructed from Durobax™ glass
  • With permanent amber graduation
  • Acid and alkali resistant 
Specifications
Autoclavable Autoclavable
Certifications/Compliance CE EN 46001
Material Borosilicate Glass
Description Amber graduation
Product Type All Glass Luer-lock Syringe
Graduations 0.2 mL
Quantity 1/Ea.
Volume (Metric) 10 mL
Connection Type Luer Lock
Graduated Yes