Learn More
Poulten Graf™ Fortuna™ Optifix™ Safety Dispenser
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
2 to 10 mL
2.0 to 10 mL
20 to 100 mL
6 to 30 mL
Includes:
PTFE-coated Piston, Stopcock and Ventilator Valve
PTFE-encased Piston, Sealing Lip
Strongly PTFE-encased Special Piston
Unit Size:
Each
Description
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
- Manufactured according to DIN EN ISO 9001:2008 using high quality materials
- With certificate, individually numbered
- Glass dispensing cylinder is coated with plastic to provide breakage protection
- Ergonomic finger protection cap made of PP with volume setting system
- Graduation on the scale is lasered and easy to read
- Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
| Product Type | Outfix Safety |
| Volume (Metric) | 2 to 10 mL |
| Includes | PTFE-coated Piston, Stopcock and Ventilator Valve |