Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
13.0 cu. ft. Freezers
17.3 cu. ft. Freezers
23.0 cu. ft. Freezers
28 cu. ft. Freezers
Type:
Shelf Kit
Unit Size:
Each
Beskrivelse
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
- Stainless steel
- Kits include mounting hardware
Tekniske data
Tekniske data
| Type | Shelf Kit |
| Capacity (English) | 23 cu. ft. |
| Capacity (Metric) | 651.2 L |
| Style | Upright |
| Color | Silver |
| For Use With (Equipment) | 23.0 cu. ft. Freezers |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion