Learn More
WLD Tec™ Foot pedal 3m cable plastic Fuego SCS
Click to view available options
Includes:
3 m Cable
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Fuego SCS |
| Material | Plastic |
| Includes | 3 m Cable |