Learn More
Bode Chemie™ Foliodress™ Protect Special Mask
Foliodress™ Protect Mask - The surgical face masks with optimal filter performance.
Brand:  Bode Chemie™ 9925331
Product Code. 15346921
  • € 1035.00 / Pack of 50
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Soft combination of materials
  • With sewn ribbons
  • Particularly suitable for larger face shapes and for those who wear glasses and beards
  • Type II according to EN 14683:2014.
Specifications
Specifications
EN 14683:2014
Sewn Ribbons
50/Pk.
For Larger Face Shapes and for Those Who Wear Glasses and Beards
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers