Learn More
Memmert™ Flush-Fit Unit Without Air Slots
Product Code. 17139764 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17139764
Brand: Memmert™ HPP260ecoB29739
Only null left
Add to basket
Add to basket
Stainless steel frame covering gap between oven and wall opening
  • Stainless steel frame covering gap
  • Between oven and wall opening
  • Accessory
Specifications
For Use With (Equipment) HPP260eco
Material Stainless Steel