Learn More
Fluorescein-ERM (LRRKtide)
Fluorescein-ERM (LRRKtide)
Brand: Invitrogen PV4901
Description
A fluorescein-labeled peptide substrate based upon ezrin/radixin/moesin (ERM) that is phosphorylated by LRRK2 and LRRK2 mutants.For use in combination with the LanthaScreen™ Tb-anti-ERM (pLRRKtide) Antibody (PV4899) to evalute inhibitors in medium to high throughput screening applications.

Specifications
Fluorescein-GAGRLGRDKYKTLRQIRQ | |
400 μM Fluorescein-ERM (LRRKtide) peptide substrate in water. Store at -20°C and protect from light. | |
1 mg | |
Peptide-Based Substrate | |
Serine/Threonine Kinases | |
FITC (Fluorescein) | |
Dry Ice | |
LRRK2 |
Biochemical terbium phospho-specific activity | |
Fluorescence | |
ERM (LRRKtide) | |
Kinase Substrate | |
120892 | |
2475.8 Da | |
LRRK2 D1994A, LRRK2 R1441C, LRRK2 I2020T, LRRK2 G2019S, LRRK2 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2682386B | Certificate of Analysis | 12/09/2023 | PV4901 |
2682386A | Certificate of Analysis | 28/07/2023 | PV4901 |
2422941C | Certificate of Analysis | 15/04/2023 | PV4901 |
2422941B | Certificate of Analysis | 02/12/2022 | PV4901 |
2422941A | Certificate of Analysis | 15/04/2022 | PV4901 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.