Learn More
Thermo Scientific Chemicals Fluorescein, disodium salt, pure
€ 57.01 - € 704.51
Chemical Identifiers
CAS | 518-47-8 |
---|---|
Molecular Formula | C20H10Na2O5 |
Molecular Weight (g/mol) | 376.28 |
MDL Number | MFCD00167039 |
InChI Key | NJDNXYGOVLYJHP-UHFFFAOYSA-L |
IUPAC Name | disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
SMILES | [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10387590
|
Thermo Scientific Chemicals
173241000 |
100 g | Glass Bottle |
€ 57.01
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10206090
|
Thermo Scientific Chemicals
173245000 |
500 g | Glass Bottle |
€ 201.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11946941
|
Thermo Scientific Chemicals
173240025 |
2.5 kg | Plastic drum |
€ 704.51
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
518-47-8 | |
376.28 | |
NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12Show More |
C20H10Na2O5 | |
MFCD00167039 | |
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
Specifications
1700 (at lambda max.) min. | |
518-47-8 | |
C20H10Na2O5 | |
19, V,6, 456 | |
Acid Yellow 73, Fluorescein, water soluble, Uranine, C.I. 45350 | |
NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 | |
376.28 | |
376.27 | |
Pure | |
Powder |
320.0°C | |
1.2 to 1.7 | |
MFCD00167039 | |
15, 4192 | |
Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol | |
Authentic | |
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate | |
488 to 491nm (in buffer pH 8) | |
10% (105°C) max. | |
Brown to Orange/Red or Brown/Green | |
Fluorescein, disodium salt |
Safety and Handling
Emergency Overview
Irritating to eyes. May cause skin and respiratory tract irritation.
NFPA
Health: 2
Flammability:
Warning
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.