Learn More
Molecular Probes™ Fluo-4 NW Calcium Assay Kit
Click to view available options
Quantity:
10 Microplates
100 Microplate(s)
Unit Size:
1 set
Each
Description
*The Fluo-4 NW (no-wash) Calcium Assay Kits provide a simplified method for detecting intracellular calcium, without a quencher dye or a wash step.
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Calcium Assay |
| For Use With (Equipment) | Microplate Reader |
| Product Type | Stain |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 100 Microplate(s) |
| Shipping Condition | Room Temperature |