Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Product Code. 17860687 Shop All Thermo Scientific Products
Click to view available options
Tubing Size:
10 in. TPE 374, 19.7 in. Pharmed BPT
Connector Types:
Luer
MPC
MPX
Dimensions:
1/2 in. ID x 3/4 in. OD
1/4 in. ID x 3/8 in. OD
1/4 in. ID x 7/16 in. OD
1/8 in. ID x 1/4 in. OD
3/8 in. ID x 5/8 in. OD
Unit Size:
Pack of 10
Product Code. 17860687
Brand: Thermo Scientific™ SH31114.01
Denne vare kan ikke returneres. Se returpolitik
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Tekniske data
Dimensions 1/4 in. ID x 3/8 in. OD
Tubing Style 6.35 mm (1/4 in.) ID x 9.52 mm (3/8 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25 m (10 in.); TPE 374; MPC
Connector Types MPC
Product Type Pump Set
Tubing Size 10 in. TPE 374, 19.7 in. Pharmed BPT
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.