missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FLT4 (VEGFR3) [Q890H, V1128L, H1146R], GST Tag Recombinant Protein

Product Code. p-7121751
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15418543

Brand: Invitrogen™ PR7216B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic

FLT4 (VEGFR3), a receptor tyrosine kinase, is a key regulator of embryonic blood vessel development and adult angiogenesis. This kinase has been implicated in juvenile hemangioma, kaposi sarcoma, lymphangiomas, and hereditary lymphedema. VEGFR3 has an essential role in the development of the embryonic cardiovascular system before the emergence of the lymphatic vessels.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Molecular Weight (g/mol) 85.2 kDa
Name Human FLT4 (VEGFR3) [Q890H, V1128L, H1146R], GST Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Common Name FLT4 (VEGFR3) (Q890H, V1128L, H1146R Mutant)
KinaseFamily VEGF Receptors
Protein Length 800-1297; Q890H, V1128L, H1146R
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.