Learn More
Thermo Scientific Chemicals Florisil™, 100-200 mesh
Click to view available options
Quantity:
250 g
500 g
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Particle Size | 100/120 Mesh |
Packaging | Glass Bottle |
Quantity | 500 g |
Material | Florisil |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0469087 | Certificate of Analysis | 31/01/2025 | 205445000,205442500,205440050 |
A0459656 | Certificate of Analysis | 30/01/2024 | 205445000,205442500,205440050 |
A0452096 | Certificate of Analysis | 02/05/2023 | 205445000,205442500,205440050 |
A0392682 | Certificate of Analysis | 14/12/2022 | 205445000,205442500,205440050 |
A0416330 | Certificate of Analysis | 12/12/2022 | 205445000,205442500,205440050 |
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Florisil™, 100-200 mesh >