Learn More
Scotch-Brite™ Floor Pads, Red
Click to view available options
Diameter (Metric):
203 mm
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
584 mm
Unit Size:
Pack of 10
Pack of 5
Description
- For everyday cleaning and light scrubbing
- Removes light soil, scuff marks, and black heel marks
- Cleans when damp and buffs when dry
- Conformable fibers provide maximum pad-to-floor contact
- Reversible, washable pad resists clogging and loading
- Consistent, proven performance with long floor pad life
Specifications
Specifications
| Product Type | Floor Pad |
| Color | Red |
| Diameter (Metric) | 460 mm |
| For Use With (Application) | Everyday Cleaning, Light Scrubbing |