Learn More
Scotch-Brite™ Floor Pads, Green
Product Code. 17158810
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 25
Pack of 5
Product Code. 17158810
Brand: Scotch-Brite™ 7100159885
Only null left
Add to basket
Add to basket
For the efficient, intermediate cleaning of heavily soiled floors with or without a coating. For use with single disc machines or automatic floor cleaning machines with slow speed.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pad
Color Green
Diameter (Metric) 355 mm
For Use With (Application) Scrubbing
For Use With (Equipment) Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines
Style Round