Learn More
Applied Biosystems™ FLoid™ Printer, Paper Cartridge, Ink/Paper
Product Code. 12383653
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 12383653
Brand: Applied Biosystems™ 4472351
This is a complete printer package for the FLoid™ Cell Imaging Station with all you need to immediately print your images and share or paste into your lab notebook.
This is a complete printer package for the FLoid™ Cell Imaging Station with all you need to immediately print your images and share or paste into your lab notebook. It includes a small, high quality printer, paper cartridge, ink, and paper.
This package provides you with:
  • Quality prints of your fluorescent cell images acquired on the FLoid™ Cell Imaging Station
  • Water resistant photos that last up to 100 years and are printed in just 47 seconds
  • Paper with adhesive backing so photos can be pasted directly into your lab notebook
Increase the Productivity of Your FLoid™ Cell Imaging Station
This compact photo printer is optimal for the FLoid™ cell imaging station and prints photo lab quality images of your fluorescently labeled samples in under a minute. Print beautiful, borderless credit card sized photos in vibrant colors. Sticker-based backing allows you to paste directly into your lab notebook. It is an ideal printer for the FLoid™ Cell Imaging Station that allows you to acquire three color fluorescent images in a minute. As the FLoid™ Cell Imaging Station can sit right on your lab bench, not in some distant dark room, you can image and document your results wherever you want.
The prints will be water-resistant and can last up to 100 years! You'll enjoy photo lab quality and have fun all at the same time. Preview your images on the 2.5' Tilt-up LCD and easily turn your favorite pictures into an art-inspired collage with the image Shuffle feature.
TRUSTED_SUSTAINABILITY
Specifications
Type Printer Package
Product Line Floid
For Use With (Equipment) Floid™ Cell Imaging System
Content And Storage Compact photo printer with credit card size paper cartridge, color ink and paper.
Store dry at room temperature.
Quantity 1 Kit
Shipping Condition Room Temperature