Learn More
Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes
Shop All Cole-Parmer Products
Click to view available options
Temperature (Metric):
-150°C to +100°C
-150°C to +1093°C
-150°C to +1100°C
-150°C to +148°C
-150°C to +204°C
-150°C to +427°C
-150°C to +482°C
-150°C to +649°C
-200°C to +311°C
-250°C to +1100°C
Style:
Thermocouple Type: K
Type K Com Pen TC Probe, Grnd
Type K Compact Pen TC Probe Grnd
Type K Compact TC Probe, Grnd
Type K Detach TC Probe, Pen., Grnd
Type K HT Flex TC Probe
Type K HT Probe Mini Con Exp
Type K Hose Clmp Probe Mini Con, Grd
Type K, Std Pen Probe, Mini Conn
Type K, Std Probe, Mini Conn , GRD
Type-K Comp Surf Probe Mini Conn Exp
Type-K Pipe Plug Probe NPTM Mini Con
Type-K Pipe-Ftg Probe Mini Conn
Unit Size:
Each
Description
Optimize readings and temperature accuracy for a variety of applications with these probes. Cole-Parmer™ Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes are ideal for high-temperature measurements and can be used with liquids, gases and semisolids.
All insulated-wire probes include straight insulated cable with no handle. Fiberglass and ceramic insulated probes have exposed junctions; use for high-temperature applications. Exposed junction probes are not recommended for use with aqueous solutions (type J exposed-junction probes will rust when exposed to moisture).
- Each probe comes with color coded ANSI mini-connectors: type J-black, type K-yellow, type T-blue, and type E-purple.
- Ceramic Fiber-Insulated Wire Probes for high-temperature measurements.
- General-Purpose Probes are designed for use with liquids, gases and semisolids.
- ABS handles provide a light weight design for ease of use along with durability
- Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging
Specifications
Specifications
| Certifications/Compliance | ANSI |
| For Use With (Equipment) | Temperature Meters |
| Temperature (Metric) | -150°C to +482°C |
| Type | Thermocouple Probe |
| For Use With (Application) | Permanent installation on hoses or pipes |
| Includes | 10ft. SS Braid Cable |
| Style | Type K Hose Clmp Probe Mini Con, Grd |