Learn More
Neogen Petrifilm™ Flat Spreader 6425
Product Code. 17263744
Encompass
missing translation for 'orderingAttributeHoverText'
Quantity:
2 Pk.
missing translation for 'unitSize'
Pack of 2
Product Code. 17263744
missing translation for 'mfr': Neogen Petrifilm™ 7100002605
This item requires a price update prior to being ordered.
Request a Price for this item here.
Spreader used to distribute the sample evenly for Neogen Petrifilm™ Staph Express Count Plates.
  • Flat spreader for Neogen Petrifilm™ Staph Express Count Plates.
  • Spreads inoculum over uniform area.
  • Clear acrylic material allows the technician to observe and control spread.
Specifications
For Use With (Equipment) 3 M™ Petrifilm™ Staph Express Count Plates.
Quantity 2 Pk.
Product Type Accessory