Learn More
Thermo Scientific™ Flat PCR Caps, strips of 12
Click to view available options
Color:
Natural
Product Type:
Flat Cap, 12-Cap Strip
PCR Plate Cap Strip
Unit Size:
Pack of 200
Pack of 80
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Flat PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
| Color | Natural |
| Certifications/Compliance | Certified DNA/RNase Free |
| Material | Polypropylene |
| No. of Wells | 12 |
| Quantity | 25 packs of 8 |
| For Use With (Application) | Thermal Cyclers |
| Format | 200 Strip Caps |
| For Use With (Equipment) | Thermal Cyclers |
| Product Type | PCR Plate Cap Strip |
| Certified RNase/DNase-free | Yes |
| Show More |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido