Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck and Two Angled Side Necks
Click to view available options
Height (Metric):
75 mm
85 mm
95 mm
110 mm
125 mm
150 mm
165 mm
Includes:
Laboratory Flange 100
Laboratory Flange 140
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
Specifications
Specifications
| Material | Borosilicate Glass, Duran Glass |
| For Use With (Application) | Reaction vessels |
| Height (Metric) | 75 mm |
| Includes | Laboratory Flange 60 |
| Product Type | Flange lid |