missing translation for 'onlineSavingsMsg'
Learn More
Kimble Chase™ Flat Bottom Borosilicate Glass Serum Vials Product Code.: 10460041
Estimated Shipment: 15-05-2025
to see stock.
Add to basket

Kimble Chase™ Flat Bottom Borosilicate Glass Serum Vials

Product Code. 10460041
20 mL
Click to view available options
Capacity (Metric):
4 mL
20 mL
25 mL
This item is not returnable. View return policy

Product Code. 10460041

Brand: Kimble Chase™ G50752300C0U7

Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Handle, contain and store a variety of liquids, including reagents, vaccines, blood plasma, culture media and chromatography samples with Kimble™ Flat Bottom Borosilicate Glass Serum Vials.

  • Borosilicate glass
  • Flat bottom
  • 100/Pack
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 20 mL
For Use With (Application) Ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples
Material Borosilicate Glass
No. per Pack 100Pack
Packaging 100Pack
Quantity 100Pack
Shape Flat Bottom
Height (Metric) 75 mm
Product Type Serum Vial
Diameter (Metric) 23 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimble Chase™ Flat Bottom Borosilicate Glass Serum Vials >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.