missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimble Chase™ Flat Bottom Borosilicate Glass Serum Vials
Click to view available options
Capacity (Metric):
4 mL
20 mL
25 mL
Description
- Borosilicate glass
- Flat bottom
- 100/Pack

Specifications
Specifications
Capacity (Metric) | 4 mL |
For Use With (Application) | Ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples |
Material | Borosilicate Glass |
Shape | Flat Bottom |
Height (Metric) | 45 mm |
Product Type | Serum Vial |
Diameter (Metric) | 15 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kimble Chase™ Flat Bottom Borosilicate Glass Serum Vials >
Spot an opportunity for improvement?Share a Content Correction