missing translation for 'onlineSavingsMsg'
Learn More
Kimble Chase™ Flat Bottom Amber Glass Serum Vial Product Code.: 10616751
Add to basket

Kimble Chase™ Flat Bottom Amber Glass Serum Vial

Product Code. 10616751
5 mL
Click to view available options
Capacity (Metric):
5 mL
This item is not returnable. View return policy

Product Code. 10616751

Brand: Kimble Chase™ G80382000A5U2

Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Capacity (Metric) 5 mL
Color Amber
For Use With (Application) Ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples, and more
Material Glass
No. per Pack 100Pack
Packaging 100Pack
Quantity 100Pack
Shape Flat Bottom
Height (Metric) 38 mm
Product Type Serum Vial
Diameter (Metric) 20 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimble Chase™ Flat Bottom Amber Glass Serum Vial >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.