Learn More
Grant Instruments™ Flask Platforms for use with ES-20/80
Click to view available options
Holds:
16 x 250 to 300 mL Flasks
30 x 100 to 150 mL Flasks
6 x 1000 mL Flasks
9 x 500 mL Flasks
Unit Size:
Each
Description
- Interchangeable
- Clamps securely hold flasks in place during vigorous mixing
- Optional accessories for the ES-20/80 Orbital Shaker-Incubator
Specifications
Specifications
| Holds | 30 x 100 to 150 mL Flasks |
| Dimensions (L x W x H) | 36 x 40 x 10 cm |
| Weight (English) | 7.3 lb. |
| Weight (Metric) | 3.3 kg |
| Description | Platform for 100 mL Flasks (30 x 100-150 mL Flasks. ES-20/80 and ES-20/80 C) |
| Model | HSP-30/100 |
| For Use With (Equipment) | ES-20/80 and ES-20/80C |