Learn More
Macherey-Nagel™ Flash columns RS 1600 C18
Brand:  Macherey-Nagel™ 732819
Product Code. 12672398
  • € 4800.00 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Specifications
430 mm
1760 g
Flash Column
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Flash columns RS 1600 C18 > Length: 430mm