Learn More
Liebherr™ FKvsl Series Premium Forced Air Refrigerators
Maintain a uniform temperature and consistent humidity level throughout the interior. Liebherr™ FKvsl Series Premium Forced Air Refrigerators are made of premium stainless steel, ergonomically designed and easy to clean. Available in various sizes with either a solid steel or glazed glass door.
Brand:  Liebherr™ FKvsl 2613
 View more versions of this product
Product Code. 15776828
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Dynamic forced air cooling: Cools fresh items quickly and features a door-operated fan switch that prevents warm air from the outside from entering the appliance when open
  • Stainless steel housing: Constructed of a highly robust alloy steel that is extremely hygienic and ideal for the professional sector
  • Efficient refrigeration system: Reduces energy consumption and operating costs with premium-quality compressors, condensers, fans and other components
  • Digital temperature display: Lets users easily view the interior temperature to the exact degree from the outside
  • SwingLine door with bar handle and reversible hinges: Stylishly contoured, ergonomically designed for any space, easy to clean and easy to open even with constant use
  • Seamless plastic inner container: Thermoformed one-piece interior is extremely easy to clean; robust and long-lasting; and made of environmentally friendly, recyclable polystyrene
  • Adjustable grid shelves: Robust shelves are height-adjustable so the interior layout can be varied as required for items of all sizes stored upright or laying down
  • Environmentally friendly refrigerant: Minimal greenhouse effect and no ozone depletion potential with outstanding energy efficiency
Specifications
Specifications
General Purpose Refrigerator
1°C to 15°C
Self-closing SwingLine
Right Interchangeable
45 kg
Thermoformed One-piece White Plastic
Silver
Adjustable feet
Double-glazed glass door
FKvsl 2613
61 x 60 x 125 cm
125 cm
1.0 A
220 to 240 V 50 Hz 1.0 A
1.030 kWh / 24 hr.
60 kg
250 L
R 600a
1
5 (4 Adjustable) Grid
Stainless Steel
Digital, External
Mechanical
Automatic
Ceiling
Key Lock
61 cm
60 cm
220/240 V
50 Hz
65 kg