Learn More
Kartell™ Fixed Volume Labofix Micropipettor
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
Compatible Tips:
Kartell™ 956/959
Kartell™ 958/960
Kartell™ 961
missing translation for 'unitSize'
Each
Description
- Made of high quality plastic
- With tip ejector and stainless steel piston for prolonged use over time
- Handle connection ensures a more comfortable and non-fatiguing grip
- Tip holder cone can house a wide variety of tips
- It has a fine shape to make getting inside test tubes or containers having particularly narrow openings or lengthened shapes easier
- Robust structure of the body ensures high mechanical and chemical resistances
Specifications
Specifications
| Accuracy | ±0.5% |
| Certifications/Compliance | EC/IVD, ISO8655 |
| Compatible Tips | Kartell™ 958/960 |
| Autoclavable | Autoclavable |
| Material | Plastic |
| Model | Labofix 25 |
| Type | Fixed Volume Micropipettor |
| Volume (Metric) | 25 μL |
| Precision | ±0.3% |