Learn More
SCAT™ Ferrule for HPLC Systems
Shop All SCAT Europe Products
Click to view available options
Diameter (Metric) Inner:
1.6 mm
2.3 mm
3.2 mm
Unit Size:
Pack of 10
Description
SCAT™ fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Specifications
| Type | Ferrule |
| Color | White |
| Diameter (English) Inner | 0.09 in. |
| Diameter (Metric) Inner | 2.3 mm |
| For Use With | Capillary |
| Material | PTFE |
| Autoclavable | Autoclavable |