Learn More
Fisherbrand™ Sterile Sampling Bags with Flat-Wire Closures
Product Code. 11632569 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
10 x 10 in.
12 x 7 in. (30 x 18cm)
14 x 10 in. (35.6 x 25.4cm)
15 x 5.5 in. (38.1 x 14cm)
9 x 4.5 in. (22.9 x 11.4cm)
9 x 5.5 in. (22.9 x 14cm)
Thickness (English):
3 mil
3.50 mil
4 mil
Color:
Clear
Clear with White Block
Capacity (English):
18 oz.
24 oz.
42 oz.
55 oz.
60 oz.
100 oz.
Unit Size:
Pack of 250
Pack of 500
Product Code. 11632569
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
An economical and efficient way to collect, contain and carry samples
Collect, contain and carry samples in an economical and efficient way
  • Sterile to avoid sample contamination
Specifications
Capacity (English) 60 oz.
Color Clear with White Block
Material Polyethylene
Sterility Sterile
Quantity 250/Pk.
Thickness (English) 4 mil
Dimensions (L x W) 10 x 10 in.
Length (English) 10 in.
Length (Metric) 25.4 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Product Type Sample Bag