Learn More
Fisherbrand™ Square Labels
GREENER_CHOICE
Product Code. p-4701710 Shop All Fisherbrand Products
Click to view available options
Color:
Assorted
Blue
Green
Orange
Red
White
Yellow
Product Type:
Square Label
Quantity:
156/Pk.
52/Sheet
85 Labels/Sheet
Size (Metric):
0.64 x 3.17 cm
0.64 x 3.81 cm
0.64 x 3.81cm
1.91 x 4.29 cm
Unit Size:
20 sheets
25 sheets
Product Code. 17807201
Only null left
Add to basket
Add to basket
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
  • Labels can be printed or written on and will not bleed, smudge or smear
  • Engineered for use in laser printers
Specifications
Color White
Format Sheet
Product Type Square Label
Sterility Non-sterile
Labels per Sheet 52
Quantity 52/Sheet
Size (English) .75 x 1.69 in.
Size (Metric) 1.91 x 4.29 cm