Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Shop All Fisherbrand Products
Click to view available options
Color:
Clear with Red Print
Red with Black Print
Dimensions (L x W):
12 x 8 in. (30 x 20 cm)
23 x 19 in. (58 x 48 cm)
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Thickness:
1.25 mil
1.50 mil
Unit Size:
Pack of 100
Pack of 1000
Pack of 200
Description
Strong and leak-resistant
- Printed with biohazard symbol and text in English and Spanish
- Supplied with twist tie closure
Specifications
Specifications
| Closure Type | Twist Tie |
| Color | Clear with Red Print |
| Dimensions (L x W) | 24 x 12 in. (61 x 30 cm) |
| Length (Metric) | 61 cm |
| Width (Metric) | 30 cm |
| Autoclavable | Autoclavable |
| Includes | Biohazard symbol and text in English and Spanish |
| Width (English) | 12 in. |
| Length (English) | 24 in. |
| Thickness | 1.50 mil |
| Show More |