Learn More
Fisherbrand™ Mild Steel Bow Tongs
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
200 mm
410 mm
Unit Size:
Each
Spécification
Spécification
| Includes | Bow and Flat Joint |
| Quantity | 1 Ea. |
| Length (Metric) | 200 mm |
| Material | Steel |
| Type | Crucible Tongs |