Learn More
Fisherbrand™ Round Bottom Disposable Borosilicate Glass Tubes with Plain End
Product Code. p-192824 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
4 mL
6 mL
10 mL
13 mL
15 mL
19 mL
23 mL
28 mL
36 mL
55 mL
Length (Metric):
75 mm
85 mm
100 mm
125 mm
150 mm
Diameter (Metric):
10 mm
12 mm
13 mm
15 mm
16 mm
18 mm
20 mm
25 mm
Unit Size:
Pack of 1000
Pack of 500
Product Code. 12347279
Only null left
Add to basket
Add to basket
Use these tubes for tissue culture and clinical chemistry applications. Fisherbrand™ Disposable Borosilicate Glass Tubes with Plain End are made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass.
Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass. These disposable borosilicate glass tubes are perfect for tissue culture and clinical chemistry applications.
  • 10 and 12mm O.D. sizes are available
  • Packaged in shrink-wrapped trays of 125 or 250 to prevent contamination or breakage
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Design Type Plain
Capacity (Metric) 4 mL
Length (Metric) 75 mm
Material Borosilicate Glass
Disposable Single-use
For Use With For tissue culture and clinical chemistry
Packaging Shrink Wrapped Tray
Rimmed Yes
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Diameter (Metric) 10 mm
Closure Type KIM-KAP
Product Type Disposable Culture Tube