Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
Click to view available options
Thickness (Metric) Teeth:
0.05 cm
0.08 cm
0.15 cm
0.77 cm
Teeth Count:
6
8
10
12
15
20
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Owl P8 Series Electrophoresis System |
| Type | Comb |
| Gel Dimensions | 10 x 10 cm |
| Teeth Count | 8 |
| Thickness (Metric) Teeth | 0.15 cm |
| Volume (Metric) Well | 185 μL |
| Length (English) Footprint | 5.9 in. |
| Length (English) Gel | 3.9 in. |
| Length (Metric) Footprint | 15 cm |
| Length (Metric) Gel | 10 cm |
| Show More |