Learn More
Fisherbrand™ Combs for FB-VE16-1 Vertical Electrophoresis System
Click to view available options
Volume (Metric) Well:
51 μL
69 μL
96 μL
107 μL
129 μL
183 μL
201 μL
343 μL
Length (Metric) Footprint:
16 cm
20 cm
Teeth Count:
10
15
20
24
Width (Metric) Teeth:
0.29 cm
0.39 cm
0.61 cm
1.04 cm
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Owl P1-CST Multiple Gradient Gel Caster |
| Type | Comb |
| Gel Dimensions | 16 x 14 cm |
| Teeth Count | 24 |
| Thickness (Metric) Teeth | 0.08 cm |
| Volume (Metric) Well | 51 μL |
| Length (English) Footprint | 7.87 in. |
| Length (English) Gel | 6.3 in. |
| Length (Metric) Footprint | 20 cm |
| Length (Metric) Gel | 16 cm |
| Show More |