Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Spare Bottles
Product Code. 11707996 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
2 L
4 L
Unit Size:
Each
Product Code. 11707996
Brand: Thermo Scientific™ N10821
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Subsitute bottles of different sizes for added convenience
ELISA-based assays, cell washing
TRUSTED_SUSTAINABILITY
Specifications
Includes Solid cap
Capacity (English) 67.62 oz.
Capacity (Metric) 2 L
For Use With (Equipment) Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer