missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Refrigerator/Freezer Thermometer

Product Code. p-2371227 Shop All Fisherbrand Products
Click to view available options
Accuracy:
±0.5°C
Resolution:
0.1°C
Unit Size:
Each

Product Code. 10501255

Only null left
Add to basket
Add to basket

Insulated sensors protect against rapid temperature changes. Fisherbrand Refrigerator/Freezer Thermometers accurately monitor temperatures in freezers, water baths, heating blocks, incubators and refrigerators with bottle-enclosed temperature buffered sensor.

  • Nontoxic glycol solution in bottle insulates sensor from rapid temperature changes when door is open
  • Triple display shows high, low and current temperatures, and min/max feature monitors high and low readings for any time period
  • Alarm feature provides an alert when temperature rises above or falls below a setpoint
  • 10 ft. micro-cable permits refrigerator doors to close on it
  • Velcro™ fasteners and magnetic strips are provided for mounting of both thermometer and bottle sensor
  • Supplied with one-year AA battery

S66277

  • Range: -50° to +70°C (-58° to +158°F)
  • Resolution: 1°
  • Accuracy: ±1°C

S66278

  • High-resolution, high-accuracy and min/max memory
  • Range: -50° to +70°C (-58° to +158°F)
  • Resolution: 0.1°
  • Accuracy: ±0.5°C

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Certifications/Compliance Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Dimensions (L x W x H) 3.25 x 3.25 in. (8.3 x 8.2 cm)
For Use With (Application) Monitoring liquids, air/gas or semi solids in freezers, water baths, outdoor tanks, incubators and refrigerators
Includes AAA battery, Velcro™ and magnetic strips are provided for mounting of both thermometer and bottle sensor
Length (English) Overall 3.25 in.
Length (Metric) 8.26 cm
Length (Metric) Overall 8.3 cm
Material Plastic
Resolution 0.1°C
Weight (English) 4 oz.
Weight (Metric) 110 g
Width (English) 3.25 in.
Product Type Refrigerator/Freezer Thermometer
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
For Use With (Equipment) Refrigerators/Freezers
Accuracy ±0.5°C
Display Digital LCD
Width (Metric) 8.2 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.