Learn More
Fisherbrand™ Refrigerator/Freezer Thermometer
Insulated sensors protect against rapid temperature changes. Fisherbrand Refrigerator/Freezer Thermometers accurately monitor temperatures in freezers, water baths, heating blocks, incubators and refrigerators with bottle-enclosed temperature buffered sensor.
Brand: Fisherbrand™ FB 50268
50.03 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Nontoxic glycol solution in bottle insulates sensor from rapid temperature changes when door is open
- Triple display shows high, low and current temperatures, and min/max feature monitors high and low readings for any time period
- Alarm feature provides an alert when temperature rises above or falls below a setpoint
- 10 ft. micro-cable permits refrigerator doors to close on it
- Velcro™ fasteners and magnetic strips are provided for mounting of both thermometer and bottle sensor
- Supplied with one-year AA battery
S66277
- Range: -50° to +70°C (-58° to +158°F)
- Resolution: 1°
- Accuracy: ±1°C
S66278
- High-resolution, high-accuracy and min/max memory
- Range: -50° to +70°C (-58° to +158°F)
- Resolution: 0.1°
- Accuracy: ±0.5°C
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™. | |
Monitoring liquids, air/gas or semi solids in freezers, water baths, outdoor tanks, incubators and refrigerators | |
3.25 in. | |
8.3 cm | |
0.1°C | |
110 g | |
Refrigerator/Freezer Thermometer | |
70°C | |
-50°C | |
-50°C to +70°C | |
Refrigerators/Freezers | |
Digital LCD |
3.25 x 3.25 in. (8.3 x 8.2 cm) | |
AAA battery, Velcro™ and magnetic strips are provided for mounting of both thermometer and bottle sensor | |
8.26 cm | |
Plastic | |
4 oz. | |
3.25 in. | |
158°F | |
-58°F | |
-58°F to +158°F | |
Yes | |
±0.5°C | |
8.2 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.